Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045903.1 | NP_001039368.1 | 3 - 4 | 542 | 126 | |
GenBank mRNA | BC113221.1 | - | 3 - 4 | 542 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |