Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046115.2 | NP_001039580.1 | 1 - 2 | 132 | 66 | |
GenBank mRNA | BC105332.1 | - | 1 - 2 | 133 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |