Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046218.1 | NP_001039683.1 | 8 - 9 | 1189 | 79 | |
GenBank mRNA | BC123801.1 | - | 8 - 9 | 1173 | 79 | |
DQ432067.2 | - | 8 - 9 | 1189 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |