Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046618.1 | NP_001040083.1 | 1 - 2 | 74 | 75 | |
GenBank mRNA | BC114865.1 | - | 1 - 2 | 74 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |