Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075967.1 | NP_001069435.1 | 4 - 5 | 351 | 93 | |
GenBank mRNA | BC114858.1 | - | 4 - 5 | 351 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |