Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077112.1 | NP_001070580.1 | 4 - 5 | 322 | 79 | |
GenBank mRNA | BC116161.1 | - | 4 - 5 | 322 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |