Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075335.1 | NP_001068803.1 | 5 - 6 | 957 | 69 | |
GenBank mRNA | BC119939.1 | - | 5 - 6 | 957 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |