Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075551.1 | NP_001069019.1 | 2 - 3 | 248 | 113 | |
GenBank mRNA | BC126750.1 | - | 2 - 3 | 239 | 113 | |
DQ435444.1 | - | 2 - 3 | 248 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |