Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079646.1 | NP_001073114.1 | 4 - 5 | 162 | 92 | |
GenBank mRNA | BC126739.1 | - | 4 - 5 | 162 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |