Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101049.2 | NP_001094519.2 | 9 - 10 | 1167 | 107 | |
GenBank mRNA | BC149742.1 | - | 5 - 6 | 610 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |