Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024533.2 | NP_001019704.2 | 1 - 2 | 89 | 132 | |
GenBank mRNA | BC119980.1 | - | 1 - 2 | 89 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |