Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098878.1 | NP_001092348.1 | 3 - 4 | 557 | 110 | |
GenBank mRNA | BC146174.1 | - | 3 - 4 | 557 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |