Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098911.2 | NP_001092381.1 | 3 - 4 | 576 | 82 | |
GenBank mRNA | BC142410.1 | - | 3 - 4 | 577 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |