Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098983.1 | NP_001092453.1 | 2 - 3 | 538 | 73 | |
GenBank mRNA | BC110237.1 | - | 2 - 3 | 533 | 73 | |
BC146164.1 | - | 2 - 3 | 538 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |