Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104970.1 | NP_001098440.1 | 2 - 3 | 306 | 124 | |
GenBank mRNA | BC149059.1 | - | 2 - 3 | 306 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |