Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105331.2 | NP_001098801.2 | 5 - 6 | 688 | 103 | |
NM_001164088.1 | NP_001157560.1 | 5 - 6 | 703 | 103 | ||
GenBank mRNA | BC153238.1 | - | 5 - 6 | 689 | 103 | |
BT026215.1 | - | 5 - 6 | 654 | 103 | ||
BT026330.1 | - | 5 - 6 | 693 | 103 | ||
BT029825.1 | - | 5 - 6 | 691 | 103 | ||
BT029828.1 | - | 5 - 6 | 670 | 103 | ||
CR452506.1 | - | 2 - 3 | 234 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |