Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARVELD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099055.1 | NP_001092525.1 | 2 - 3 | 524 | 85 | |
GenBank mRNA | BC146084.1 | - | 2 - 3 | 524 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |