Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099065.2 | NP_001092535.1 | 4 - 5 | 529 | 65 | |
GenBank mRNA | BC142453.1 | - | 4 - 5 | 531 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |