Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105426.1 | NP_001098896.1 | 4 - 5 | 432 | 81 | |
GenBank mRNA | BC134656.1 | - | 4 - 5 | 432 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |