Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP11L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105431.1 | NP_001098901.1 | 8 - 9 | 1273 | 122 | |
GenBank mRNA | BC153231.1 | - | 8 - 9 | 1273 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |