Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111104.1 | NP_001104574.1 | 4 - 5 | 845 | 78 | |
GenBank mRNA | BC134633.1 | - | 4 - 5 | 845 | 78 | |
EE359881.1 | - | 3 - 4 | 402 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |