Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113301.1 | NP_001106772.1 | 1 - 2 | 138 | 56 | |
GenBank mRNA | BC108230.1 | - | 1 - 2 | 138 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |