Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101881.2 | NP_001095351.2 | 4 - 5 | 655 | 104 | |
GenBank mRNA | BC102355.1 | - | 3 - 4 | 555 | 104 | |
BC149884.1 | - | 3 - 4 | 555 | 104 | ||
EV674476.1 | - | 3 - 4 | 612 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |