Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163458.1 | NP_001156930.1 | 11 - 12 | 2018 | 70 | |
GenBank mRNA | BC126681.1 | - | 11 - 12 | 1958 | 70 | |
DV911405.1 | - | 3 - 4 | 298 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |