Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205464.1 | NP_001192393.1 | 1 - 2 | 87 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |