Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VANGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205875.1 | NP_001192804.1 | 6 - 7 | 1514 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |