Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001354198.1 | NP_001341127.1 | 3 - 4 | 1095 | 108 | |
GenBank mRNA | EE241826.1 | - | 1 - 2 | 273 | 108 | |
EH139359.1 | - | 2 - 3 | 433 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Probe spans exons |