Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CILP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192528.1 | NP_001179457.1 | 6 - 7 | 1041 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |