Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192567.1 | NP_001179496.1 | 4 - 5 | 877 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |