Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192649.1 | NP_001179578.1 | 3 - 4 | 1154 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |