Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206295.1 | NP_001193224.1 | 4 - 5 | 685 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |