Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206564.1 | NP_001193493.1 | 4 - 5 | 308 | 82 | |
GenBank mRNA | BC110167.1 | - | 4 - 5 | 574 | 82 | |
DV869571.1 | - | 4 - 5 | 294 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |