Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192672.3 | NP_001179601.1 | 5 - 6 | 1482 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |