Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4K2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192769.1 | NP_001179698.1 | 2 - 3 | 247 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |