Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206597.1 | NP_001193526.1 | 6 - 7 | 858 | 92 | |
GenBank mRNA | DV916511.1 | - | 4 - 5 | 548 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |