Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206623.1 | NP_001193552.1 | 3 - 4 | 725 | 62 | |
GenBank mRNA | CR848913.2 | - | 3 - 4 | 449 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |