Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206764.2 | NP_001193693.1 | 3 - 4 | 316 | 77 | |
GenBank mRNA | EE353771.1 | - | 2 - 3 | 241 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |