Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206982.1 | NP_001193911.1 | 3 - 4 | 507 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |