Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245934.1 | NP_001232863.1 | 2 | 524 | 104 | |
GenBank mRNA | HM136903.1 | - | 2 | 524 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |