Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hch-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078039.7 | NP_510440.1 | 1 - 2 | 61 | 70 | |
GenBank mRNA | D85744.1 | - | 1 - 2 | 162 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |