Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001364051.5 | NP_001350994.1 | 1 | 197 | 79 | |
NM_001364052.5 | NP_001350995.1 | 1 | 197 | 79 | ||
NM_001364053.4 | NP_001350996.1 | 3 - 4 | 212 | 79 | ||
NM_001364054.4 | NP_001350997.1 | 3 - 4 | 212 | 79 | ||
NM_001368560.4 | NP_001355485.1 | 1 | 197 | 79 | ||
NM_078144.4 | NP_510545.1 | 3 - 4 | 469 | 79 | ||
NM_078145.5 | NP_510546.1 | 3 - 4 | 502 | 79 | ||
NM_078146.5 | NP_510547.1 | 3 - 4 | 212 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |