Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377804.1 | NP_001364770.1 | 1 - 2 | 195 | 74 | |
NM_001381436.2 | NP_001367838.1 | 1 - 2 | 183 | 74 | ||
NM_170830.6 | NP_740814.2 | 1 - 2 | 182 | 74 | ||
NR_131363.1 | 1 - 2 | 153 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |