Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170854.5 | NP_740841.1 | 9 - 10 | 2220 | 77 | |
NM_170855.6 | NP_740842.1 | 9 - 10 | 2302 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |