Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noah-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263093.4 | NP_001250022.1 | 4 - 5 | 1502 | 93 | |
NM_170870.8 | NP_740858.2 | 4 - 5 | 1498 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |