Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171250.5 | NP_741298.2 | 2 - 3 | 428 | 72 | |
NM_171897.7 | NP_741299.2 | 2 - 3 | 402 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |