Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025921.4 | NP_001021092.1 | 3 - 4 | 1099 | 68 | |
NM_001025922.5 | NP_001021093.1 | 3 - 4 | 415 | 68 | ||
GenBank mRNA | U04711.1 | - | 3 - 4 | 418 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |