Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttx-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128981.6 | NP_001122453.2 | 4 - 5 | 476 | 97 | |
GenBank mRNA | AB303038.1 | - | 4 - 5 | 476 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |