Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059667.7 | NP_492068.1 | 1 - 2 | 377 | 96 | |
GenBank mRNA | U59210.1 | - | 1 - 2 | 354 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |