Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059712.6 | NP_492113.2 | 8 - 9 | 3702 | 121 | |
GenBank mRNA | AY874871.1 | - | 8 - 9 | 3665 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |