Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 14 - 15 | 2596 | 87 | |
NM_001026218.5 | NP_001021389.1 | 13 - 14 | 2224 | 87 | ||
NM_001026219.5 | NP_001021390.1 | 14 - 15 | 2547 | 87 | ||
NM_001128984.4 | NP_001122456.2 | 16 - 17 | 2763 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |